Learn More
Thermo Scientific Chemicals Moclobemide
Supplier: Thermo Scientific Chemicals 465802500
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
| Moclobemide | |
| 250 mg | |
| C13H17ClN2O2 | |
| YHXISWVBGDMDLQ-UHFFFAOYSA-N | |
| 4-chloro-N-[2-(morpholin-4-yl)ethyl]benzamide |
| 71320-77-9 | |
| 135.0°C to 138.0°C | |
| 268.74 | |
| ClC1=CC=C(C=C1)C(=O)NCCN1CCOCC1 | |
| 268.74 |
Chemical Identifiers
| 71320-77-9 | |
| C13H17ClN2O2 |
Safety and Handling
GHS H Statement Harmful if swallowed.
Causes skin irritation.
Causes serious eye damage.
May cause respiratory irritation.
GHS P Statement Wear protective gloves/protective clothing/eye protection/face protection.
Immediately call a POISON CENTER or doctor/physician.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
IF ON SKIN: Wash with plenty of soap and water.
GHS Signal Word: Danger
RUO – Research Use Only