missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mixed Calibration Standard 3 For Environmental Analysis, SPEX CertiPrep™
Description
Comprend
NIST Traceable Certificate of Analysis accredited to ISO/IEC 17025 and ISO/IEC Guide 34, Safety Data Sheet
Inorganic Standard manufactured for AA, ICP, ICP/MS, IC, XRF, and other analytical instrumentation
| Analyte | Concentration |
|---|---|
| As | 500μg/mL |
| Mo | 100μg/mL |
| Si | 100μg/mL |
Certifications
Accredited by A2LA for ISO/IEC 17025 and ISO/IEC Guide 34 and certified by UL-DQS to ISO 9001
Compliance
GHS/OSHA Complaint
Spécifications
Spécifications
| Chemical Name or Material | ICP Mixed Calibration Standard 3 |
| Packaging | Triple-leached LDPE bottle |
| Concentration or Composition Notes | Up to 68 elements are scanned with found values for ICP and ICP-MS standards. The found value of the trace metallic impurities is reported on the Certificate of Analysis. |
| Technique | AA, ICP |
| For Use With (Equipment) | AA, ICP, ICP/MS, IC, XRF, and Other Analytical Instrumentation |
| Type | Certified Reference Material |
| Solvent or Matrix | Nitric Acid/Trace Hydrogen Fluoride |
| Quantity | 125 mL |
| Grade | Environmental |
| Concentration or Composition (by Analyte or Components) | Components in 2% HNO3/tr. HF: As •500μg/mL Mo •100μg/mL Si •100μg/mL |
Product intended for laboratory use only