Learn More
Honeywell™ Miller™ DuraFlex™ Python™ Harnesses
Catalog No. 19274634
Change view
Click to view available options
Buckle Style:
Mating
D-Ring Location:
Back
Catalog No. Buckle Style D-Ring Location
19274634 Mating Back
1 options
Catalog No. 19274634 Supplier Honeywell™ Supplier No. P950/UGN
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Combine stretchable DuraFlex torso straps with lightweight, cushioned shoulder straps for ultimate comfort
Come with an invisible protective coating that resists staining, repels dirt and grime, preserves color, and wipes clean in an instant, ensuring easy maintenance and longer service life.
  • Abrasion-resistant, green nylon webbing has contoured tubular edges and high-density foam cushioning
  • Does not bunch or kink, improving neck and shoulder comfort
  • Flexibility and ease of movement are enhanced by stretchable webbing at the torso, which reduces fatigue
  • Adjustable leg straps feature nonstretch webbing with a choice of tongue or mating buckles
  • Fully adjustable, tangle-free, and easy to don
  • All models feature a sliding back D-ring and pad
  • Universal size
  • Green
Compliance
Meets OSHA 1910.66 and OSHA 1926.502 (d); ANSI™ A10.32 and ANSI Z359-2007; CSA Z259.10 requirements.
Specifications
Material Nylon Webbing
Color Green
Buckle Style (Chest) Mating Chest Straps
Buckle Style Mating
D-Ring Location Back
Certifications/Compliance Meets all applicable OSHA, ANSI, and CSA requirements.
Buckle Style (Leg) Mating Leg Straps