missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Integra™ Miltex™ Mid Grade Dressing Forceps
Supplier: Integra™ Miltex™ V9616
Description
- Stainless steel construction
Identifiants chimiques
| Serrated | |
| Forceps, Dressing | |
| Mid |
| Stainless Steel | |
| Each |
Spécifications
| Serrated | |
| Stainless Steel | |
| Forceps, Dressing | |
| Mid |
| 10 in. | |
| 254 mm | |
| Each |