Learn More
Eppendorf™ Assay/Reader Microplate
Eppendorf Assay/Reader Microplates are designed for diverse microplate assays, optimizing absorbance, fluorescence, and chemiluminescence. With options for both top- and bottom-reading plate readers, they ensure clear signals even with low-concentration samples.
$1053.37 - $1820.63
Specifications
| Autoclavable | Autoclavable |
|---|---|
| Max. RCF | 6000 x g |
| Purity or Quality Grade | PCR Clean (Non-sterile) |
| Catalog Number | Mfr. No. | Color | Well Shape | No. of Wells | Well Color | Price | Quantity | ||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Color | Well Shape | No. of Wells | Well Color | Price | Quantity | ||
|
05414244
|
Eppendorf™
951040196 |
White | Flat Bottom (F-well) | 96 |
Case of 80 Each for $1,053.37
|
|
|||
|
05414243
|
Eppendorf™
951040137 |
Gray | Flat Bottom (F-well) | 96 |
Case of 80 Each for $1,057.79
|
|
|||
|
E951040308
|
Eppendorf™
951040308 |
Gray | Conical Bottom (V-well) | 96 | White |
Each for $1,057.79
|
|
||
|
E951040102
|
Eppendorf™
951040102 |
White | Round Bottom (U-well) | 96 | Black |
Each for $1,144.54
|
|
||
|
E951040145
|
Eppendorf™
951040145 |
Gray | Round Bottom (U-well) | 96 | White |
Each for $1,057.79
|
|
||
|
E951040260
|
Eppendorf™
951040260 |
White | Conical Bottom (V-well) | 96 | Black |
Each for $1,144.54
|
|
||
|
E951040481
|
Eppendorf™
951040481 |
White | Conical Bottom (V-well) | 384 | Black |
Each for $1,820.63
|
|
||
|
E951040503
|
Eppendorf™
951040503 |
Gray | Conical Bottom (V-well) | 384 | White |
Each for $1,820.63
|
|
||
Description
- Black Eppendorf Microplates offer an excellent signal-to-noise ratio – for clear signals even with low-concentration samples.
- White Eppendorf Microplates are optimized for highest sensitivity in the detection of luminescence signals by maximizing reflection.
- Solid black and white assay plates are made of polypropylene resulting in high resistance to chemicals, mechanical stress, and temperature extremes.
- All plates are optimized for minimal autofluorescence and autoluminescence.
- Available in lot-certified PCR clean purity grade.
- Outstanding dimensional accuracy and high well-to well homogeneity for perfect automation suitability.
Specifications
| Autoclavable | |
| 6000 x g | |
| PCR Clean (Non-sterile) |