missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Eppendorf™ Assay/Reader Microplate
Eppendorf Assay/Reader Microplates are designed for diverse microplate assays, optimizing absorbance, fluorescence, and chemiluminescence. With options for both top- and bottom-reading plate readers, they ensure clear signals even with low-concentration samples.
Supplier: Eppendorf™ 951040481
Description
- Black Eppendorf Microplates offer an excellent signal-to-noise ratio – for clear signals even with low-concentration samples.
- White Eppendorf Microplates are optimized for highest sensitivity in the detection of luminescence signals by maximizing reflection.
- Solid black and white assay plates are made of polypropylene resulting in high resistance to chemicals, mechanical stress, and temperature extremes.
- All plates are optimized for minimal autofluorescence and autoluminescence.
- Available in lot-certified PCR clean purity grade.
- Outstanding dimensional accuracy and high well-to well homogeneity for perfect automation suitability.
Compliance
Manufactured in ISO certified clean room conditions. Each batch is tested and certified individually.
Specifications
| ISO | |
| 16 x 24 | |
| White | |
| Polypropylene | |
| 0.005 to 0.15 μL | |
| Conical Bottom (V-well) | |
| 6000 x g | |
| Black |
| Standard | |
| Autoclavable | |
| Microplate | |
| 384 | |
| Non-sterile | |
| Storing and analyzing samples | |
| PCR Clean (Non-sterile) |