missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Eppendorf™ Assay/Reader Microplate
Eppendorf Assay/Reader Microplates are designed for diverse microplate assays, optimizing absorbance, fluorescence, and chemiluminescence. With options for both top- and bottom-reading plate readers, they ensure clear signals even with low-concentration samples.
Supplier: Eppendorf™ 951040308
Description
- Black Eppendorf Microplates offer an excellent signal-to-noise ratio – for clear signals even with low-concentration samples.
- White Eppendorf Microplates are optimized for highest sensitivity in the detection of luminescence signals by maximizing reflection.
- Solid black and white assay plates are made of polypropylene resulting in high resistance to chemicals, mechanical stress, and temperature extremes.
- All plates are optimized for minimal autofluorescence and autoluminescence.
- Available in lot-certified PCR clean purity grade.
- Outstanding dimensional accuracy and high well-to well homogeneity for perfect automation suitability.
Compliance
Compliant to SBS specifications
Specifications
| SBS | |
| 8 x 12 | |
| 5 Bags x 16 Plates | |
| Microplate | |
| 350 μL | |
| 20 to 320 μL | |
| Conical Bottom (V-well) | |
| 6000 x g | |
| 80 Pk. | |
| 12.77 cm |
| Standard | |
| Autoclavable | |
| Gray | |
| Polypropylene | |
| 96 | |
| Non-sterile | |
| Fluorescence detection | |
| PCR Clean (Non-sterile) | |
| White | |
| 1.44 cm |