missing translation for 'onlineSavingsMsg'
Learn More

Eppendorf™ Assay/Reader Microplate

Eppendorf Assay/Reader Microplates are designed for diverse microplate assays, optimizing absorbance, fluorescence, and chemiluminescence. With options for both top- and bottom-reading plate readers, they ensure clear signals even with low-concentration samples.

Supplier:  Eppendorf™ 951040260

 View more versions of this product

Catalog No. E951040260

Only null left
Explore available promotions

Description

Description

  • Black Eppendorf Microplates offer an excellent signal-to-noise ratio – for clear signals even with low-concentration samples.
  • White Eppendorf Microplates are optimized for highest sensitivity in the detection of luminescence signals by maximizing reflection.
  • Solid black and white assay plates are made of polypropylene resulting in high resistance to chemicals, mechanical stress, and temperature extremes.
  • All plates are optimized for minimal autofluorescence and autoluminescence.
  • Available in lot-certified PCR clean purity grade.
  • Outstanding dimensional accuracy and high well-to well homogeneity for perfect automation suitability.

Compliance

Compliant to SBS specifications

Specifications

Specifications

SBS
8 x 12
5 Bags x 16 Plates
Microplate
350 μL
20 to 320 μL
Conical Bottom (V-well)
6000 x g
80 Pk.
12.77 cm
Standard
Autoclavable
White
Polypropylene
96
Non-sterile
Fluorescence detection
PCR Clean (Non-sterile)
Black
1.44 cm
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions