Learn More
DWK Life Sciences MicroLiter 9mm Screwthread Autosampler Vial, Cap and Septa Unassembled Component Kits with PTFE Septa
Unassembled component kits offer the convenience of one product number for the vial, cap, and septa. Components are packaged to work with the unique MicroLiter Dispenser.
Supplier:  DWK Life Sciences 091210N
 Afficher plus de versions de ce produit
Numéro de catalogue. 15111236
Il en reste null
Description
Description
Available in multiple colors and septa formats and with or without writing patch.
Spécifications
Identifiants chimiques
PTFE
Borosilicate Glass (Type 1)
2 mL
100
Screw Cap
32 mm
12 mm
No
PTFE
Polypropylene
9 mm
Spécifications
Natural
12 mm
No
PTFE
Polypropylene
9 mm
32 mm
PTFE
Borosilicate Glass (Type 1)
2 mL
100
Screw Cap
Yes
missing translation for 'recommendations'
missing translation for 'recommendations'
Videos
FDS