missing translation for 'onlineSavingsMsg'
Learn More
Learn More
International Enviroguard™ MicroGuard MP Microporous Boot Cover with Elastic Closure
Using state-of-the-art lamination technology, MicroGuard MP microporous boot covers combine a soft, non-woven fabric with a microporous film.
Supplier: International Enviroguard™ IEG8102
Description
- Lightweight, tear-resistant liquid and particulate barrier helps protect against blood penetration, various industrial chemicals, along with dust, dirt, and grime
- Breathable fabric allows moisture vapor to escape, so your feet are more cool and comfortable even as you sweat
- Generous 17 in. length provides better coverage of the feet and lower legs
- Boot covers come with a strong elastic top to help keep them in place throughout your workday
- Environmentally friendly; can be recycled after use
- Passes ASTM F1670 testing for blood penetration
Specifications
| Large | |
| Yes | |
| Single-use | |
| Elastic Top | |
| Serged | |
| 43.2 cm | |
| Boot Covers |
| Multi-layer Barrier Laminate Fabric, Spunbond Polypropylene | |
| White | |
| Non-sterile | |
| Standard | |
| 17 in. | |
| Static, Abrasion, Liquid, Grime, Dry Particulates |