missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ansell AlphaTec™ 2000 Standard Lab Coats, Bound
Low-linting lab coats provide a protective barrier to low concentration liquid chemicals and liquid and particulate biological hazards.
Supplier: Ansell WH20B9220904
Description
- Protective barrier to low concentration liquid chemicals, liquid and particulate biological hazards
- Comfortable to wear featuring moisture vapor permeable fabric (breathable) to help reduce the risk of heat stress
- Optimized fit, improving wearer comfort and safety
- Low Linting
- Free of natural rubber latex
Specifications
| Chemical Resistant Lab Coat | |
| Unisex | |
| White | |
| Elastic | |
| Non-sterile | |
| Standard Length | |
| Yes | |
| Bound | |
| CE CATEGORY III (Type PB (6)-B, EN 14126) | |
| Individually Packaged |
| Large | |
| Microporous Polyethylene Laminate | |
| Zipper | |
| Limited-use | |
| Chemicals | |
| 2 | |
| 209 | |
| ASTM F1670, ASTM F1671 | |
| Food Processing, Metal Fabrication, Cleanroom Cleaning and Preparing, Production Line Support and Maintenance, Veterinary | |
| Not Autoclavable |