missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Micro Particles Based on Melamine Resin, Analytical Standard, Size: 8 m, MilliporeSigma™ Supelco™
Spécifications
Spécifications
| Physical Form | Particles |
| Concentration | 10% (solids) |
| MDL Number | MFCD00197912 |
| Synonym | Melamine-formaldehyde resin; Size standards |
| Quantity | 5 mL |
| Grade | Analytical Standard |