missing translation for 'onlineSavingsMsg'
Learn More

Methylene Bromide, 99%, Spectrum™ Chemical

Catalog No. 18611786
Change view
Click to view available options
Quantity:
400 kg
Packaging:
Drum
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18611786 400 kg Drum
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 18611786 Supplier Spectrum Chemical Mfg Cor Supplier No. M2259400KGBL

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 74-95-3

Specifications

CAS Max % 1
Boiling Point 96.95°C/206.5°F
Packaging Drum
Quantity 400 kg
Molecular Weight (g/mol) 173.85
Percent Purity 0.99
Grade Reagent
Physical Form Liquid
Chemical Name or Material Methylene Bromide

CAUTION: For manufacturing or laboratory use only. Read and understand the label and Safety Data Sheet (SDS) prior to use.