missing translation for 'onlineSavingsMsg'
Learn More

Methylene Blue, BAKER ANALYZED™ Reagent, J.T. Baker™

High quality chemicals for laboratory and specialized industrial use

$300.15 - $5,964.65

Chemical Identifiers

CAS 7220-79-3
Molecular Formula C16H24ClN3O3S
Molecular Weight (g/mol) 373.90
MDL Number MFCD00150008
InChI Key XQAXGZLFSSPBMK-UHFFFAOYSA-M
Synonym C.I. 52015
PubChem CID 104827
SMILES O.O.O.[Cl-].CN(C)C1=CC2=[S+]C3=CC(=CC=C3N=C2C=C1)N(C)C
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Packaging Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Packaging Price Quantity & Availability  
2003392
View Documents Promotion Details
Avantor J.T.Baker
Q47303
25 g Wide Mouth Amber Glass Bottle
Each for $300.15
Only null left
 
2003391
View Documents Promotion Details
Avantor J.T.Baker
Q47305
100 g Wide Mouth Amber Glass Bottle
Case of 4 Each for $2,595.50
Only null left
 
2003394
View Documents Promotion Details
Avantor J.T.Baker
Q47307
500 g Wide Mouth Amber Glass Bottle
Case of 4 Each for $5,964.65
Only null left
 
Description

Description

Specifications

Chemical Identifiers

7220-79-3
373.90
XQAXGZLFSSPBMK-UHFFFAOYSA-M
104827
C16H24ClN3O3S
MFCD00150008
C.I. 52015
O.O.O.[Cl-].CN(C)C1=CC2=[S+]C3=CC(=CC=C3N=C2C=C1)N(C)C

Specifications

7220-79-3
MFCD00150008
XQAXGZLFSSPBMK-UHFFFAOYSA-M
[7-(dimethylamino)phenothiazin-3-ylidene]-dimethylazanium;chloride;trihydrate
104827
BAKER ANALYZED™ Reagent
C16H24ClN3O3S
C.I. 52015
O.O.O.[Cl-].CN(C)C1=CC2=[S+]C3=CC(=CC=C3N=C2C=C1)N(C)C
373.90
373.91
Methylene Blue
Videos
SDS
Documents

Documents