missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Methylene Blue, BAKER ANALYZED™ Reagent, J.T. Baker™
High quality chemicals for laboratory and specialized industrial use
Supplier: Avantor Q47305
| Quantity | 100 g |
|---|---|
| Packaging | Wide Mouth Amber Glass Bottle |
Chemical Identifiers
| C16H24ClN3O3S | |
| MFCD00150008 | |
| C.I. 52015 | |
| O.O.O.[Cl-].CN(C)C1=CC2=[S+]C3=CC(=CC=C3N=C2C=C1)N(C)C |
Specifications
| Methylene Blue | |
| C16H24ClN3O3S | |
| C.I. 52015 | |
| O.O.O.[Cl-].CN(C)C1=CC2=[S+]C3=CC(=CC=C3N=C2C=C1)N(C)C | |
| 373.90 | |
| 373.91 | |
| Wide Mouth Amber Glass Bottle |
| 7220-79-3 | |
| MFCD00150008 | |
| XQAXGZLFSSPBMK-UHFFFAOYSA-M | |
| 3,7-bis(dimethylamino)-5λ⁴-phenothiazin-5-ylium trihydrate chloride | |
| 104827 | |
| BAKER ANALYZED™ Reagent | |
| 100 g |