missing translation for 'onlineSavingsMsg'
Learn More

Methylene Blue, BAKER ANALYZED™ Reagent, J.T. Baker™

Catalog No. 2003392
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
Packaging:
Wide Mouth Amber Glass Bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
2003392 25 g Wide Mouth Amber Glass Bottle
2003391 100 g Wide Mouth Amber Glass Bottle
2003394 500 g Wide Mouth Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 2003392 Supplier Avantor Supplier No. Q47303

High quality chemicals for laboratory and specialized industrial use

Chemical Identifiers

CAS 7220-79-3
Molecular Formula C16H24ClN3O3S
Molecular Weight (g/mol) 373.90
MDL Number MFCD00150008
InChI Key XQAXGZLFSSPBMK-UHFFFAOYSA-M
Synonym C.I. 52015
PubChem CID 104827
SMILES O.O.O.[Cl-].CN(C)C1=CC2=[S+]C3=CC(=CC=C3N=C2C=C1)N(C)C

Specifications

IUPAC Name [7-(dimethylamino)phenothiazin-3-ylidene]-dimethylazanium;chloride;trihydrate
Formula Weight 373.91
Grade BAKER ANALYZED™ Reagent
Packaging Wide Mouth Amber Glass Bottle
Quantity 25 g
Chemical Name or Material Methylene Blue