missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Methyl Red Indicator, 1% (w/v), Ricca Chemical
For Acid-Base Titrations
$96.40 - $411.50
Specifications
| Chemical Name or Material | Methyl Red Indicator |
|---|---|
| CAS | 845-10-3 , 7732-18-5 |
| Percent Purity | 1% W/V |
| Grade | Indicator |
| Concentration or Composition (by Analyte or Components) | 1% (w/v) |
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity & Availability | ||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity & Availability | ||||
|
50704
|
N/A | 120 mL | Natural Poly Bottle |
Each for $96.40
|
|
||||
|
507016
|
N/A | 500 mL | Natural Poly Bottle |
Each for $237.53
|
|
||||
|
507032
|
N/A | 1 L | Natural Poly Bottle |
Each for $411.50
|
|
||||