missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Methyl Red Indicator, 1% (w/v), Ricca Chemical
For Acid-Base Titrations
Supplier: Ricca Chemical Company 507032
Chemical Identifiers
| 1% W/V | |
| 1% (w/v) | |
| pink (pH 4.2) - yellow (pH 6.2) | |
| Orange to Red | |
| Methyl Red Indicator |
Specifications
| Methyl Red Indicator | |
| 1% W/V | |
| 1% (w/v) | |
| pink (pH 4.2) - yellow (pH 6.2) | |
| Orange to Red | |
| 1 L |