missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Methyl Orange, 0.05% Aqueous, pH 3.2 to 4.4 Red to Yellow, Certified, LabChem™
$40.89 - $236.86
Chemical Identifiers
| CAS | 547-58-0 , 7732-18-5 |
|---|---|
| Molecular Formula | C14H14N3NaO3S |
| Molecular Weight (g/mol) | 327.334 |
| InChI Key | STZCRXQWRGQSJD-UHFFFAOYSA-M |
| PubChem CID | 23673835 |
| IUPAC Name | sodium;4-[[4-(dimethylamino)phenyl]diazenyl]benzenesulfonate |
| SMILES | CN(C)C1=CC=C(C=C1)N=NC2=CC=C(C=C2)S(=O)(=O)[O-].[Na+] |
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity & Availability | ||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity & Availability | ||||
|
LC170007
|
N/A | 125 mL | Poly Bottle |
Each for $40.89
|
|
||||
|
LC170001
|
N/A | 500 mL | Poly Bottle |
Each for $50.79
|
|
||||
|
LC170002
|
N/A | 1 L | Poly Bottle |
Each for $82.75
|
|
||||
|
LC170004
|
N/A | 4 L | Poly Bottle |
Each for $236.86
|
|
||||
Chemical Identifiers
| C14H14N3NaO3S | |
| STZCRXQWRGQSJD-UHFFFAOYSA-M | |
| sodium;4-[[4-(dimethylamino)phenyl]diazenyl]benzenesulfonate |
Specifications
| 547-58-0 , 7732-18-5 | |
| C14H14N3NaO3S | |
| Soluble in water | |
| STZCRXQWRGQSJD-UHFFFAOYSA-M | |
| sodium;4-[[4-(dimethylamino)phenyl]diazenyl]benzenesulfonate | |
| 23673835 | |
| Certified | |
| Orange | |
| Methyl Orange, 0.05% Aqueous, pH 3.2 to 4.4 Red to Yellow |
| 99.95, 0.05 | |
| 4-NaOSO2C6H4N:NC6H4-4-N(CH3)2 | |
| Passes Test | |
| CN(C)C1=CC=C(C=C1)N=NC2=CC=C(C=C2)S(=O)(=O)[O-].[Na+] | |
| 327.334 | |
| 327.34 | |
| Carbon monoxide; Carbon dioxide; Nitrogen oxides; Sulfur compounds | |
| Liquid |
Safety and Handling
GHS H Statement
Solution is not hazardous.
GHS P Statement
If in contact with skin or eyes, rinse thoroughly with water for 15-20 minutes.
If swallowed, get medical attention.
Recommended Storage : Room Temperature