missing translation for 'onlineSavingsMsg'
Learn More

Methyl Orange, 0.05% Aqueous, pH 3.2 to 4.4 Red to Yellow, Certified, LabChem™

Catalog No. LC170007
Change view
Click to view available options
Quantity:
125 mL
500 mL
1 L
4 L
Packaging:
Poly Bottle
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
LC170007 125 mL Poly Bottle
LC170001 500 mL Poly Bottle
LC170002 1 L Poly Bottle
LC170004 4 L Poly Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. LC170007 Supplier LabChem Supplier No. LC170007

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 547-58-0 | C14H14N3NaO3S | 327.334 g/mol

Chemical Identifiers

CAS 547-58-0 , 7732-18-5
Molecular Formula C14H14N3NaO3S
Molecular Weight (g/mol) 327.334
InChI Key STZCRXQWRGQSJD-UHFFFAOYSA-M
PubChem CID 23673835
IUPAC Name sodium;4-[[4-(dimethylamino)phenyl]diazenyl]benzenesulfonate
SMILES CN(C)C1=CC=C(C=C1)N=NC2=CC=C(C=C2)S(=O)(=O)[O-].[Na+]

Specifications

CAS Max % 99.95, 0.05
Identification Passes Test
Packaging Poly Bottle
Linear Formula 4-NaOSO2C6H4N:NC6H4-4-N(CH3)2
Decomposition Information Carbon monoxide; Carbon dioxide; Nitrogen oxides; Sulfur compounds
Color Orange
pH 3.2 to 4.4
Solubility Information Soluble in water
Quantity 125 mL
Physical Form Liquid
Formula Weight 327.34
Grade Certified
Chemical Name or Material Methyl Orange, 0.05% Aqueous, pH 3.2 to 4.4 Red to Yellow
Show More Show Less
Hazard Statement GHS H Statement
Solution is not hazardous.
Precautionary Statement GHS P Statement
If in contact with skin or eyes, rinse thoroughly with water for 15-20 minutes.
If swallowed, get medical attention.
Recommended Storage Room Temperature