missing translation for 'onlineSavingsMsg'
Learn More

Methyl Orange, 0.05% Aqueous, pH 3.2 to 4.4 Red to Yellow, Certified, LabChem™

Supplier:  LabChem LC170002

Quantity 1 L
Packaging Poly Bottle

 View more versions of this product

Catalog No. LC170002


Only null left

Description

Description

Specifications

Chemical Identifiers

547-58-0 , 7732-18-5
327.334
23673835
CN(C)C1=CC=C(C=C1)N=NC2=CC=C(C=C2)S(=O)(=O)[O-].[Na+]
C14H14N3NaO3S
STZCRXQWRGQSJD-UHFFFAOYSA-M
sodium;4-[[4-(dimethylamino)phenyl]diazenyl]benzenesulfonate

Specifications

Methyl Orange
547-58-0 , 7732-18-5
C14H14N3NaO3S
Soluble in water
STZCRXQWRGQSJD-UHFFFAOYSA-M
sodium;4-[[4-(dimethylamino)phenyl]diazenyl]benzenesulfonate
23673835
Certified
Carbon monoxide; Carbon dioxide; Nitrogen oxides; Sulfur compounds
1 L
0.05% Aqueous, pH 3.2 to 4.4 Red to Yellow
99.95, 0.05
4-NaOSO2C6H4N:NC6H4-4-N(CH3)2
Passes Test
CN(C)C1=CC=C(C=C1)N=NC2=CC=C(C=C2)S(=O)(=O)[O-].[Na+]
327.334
327.34
Poly Bottle
Orange
Liquid
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
Solution is not hazardous.

GHS P Statement
If in contact with skin or eyes, rinse thoroughly with water for 15-20 minutes.
If swallowed, get medical attention.

Recommended Storage : Room Temperature

SDS
Documents

Documents

Promotions

Promotions