Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Methyl 4-Bromobutyrate 98.0+%, TCI America™
Chemical Identifiers
| CAS | 4897-84-1 |
|---|---|
| Molecular Formula | C5H9BrO2 |
| Molecular Weight (g/mol) | 181.03 |
| MDL Number | MFCD00041482 |
| InChI Key | QAWFLJGZSZIZHO-UHFFFAOYSA-N |
| PubChem CID | 107604 |
| IUPAC Name | methyl 4-bromobutanoate |
| SMILES | COC(=O)CCCBr |
Chemical Identifiers
| 4897-84-1 | |
| 181.03 | |
| QAWFLJGZSZIZHO-UHFFFAOYSA-N | |
| methyl 4-bromobutanoate |
| C5H9BrO2 | |
| MFCD00041482 | |
| 107604 | |
| COC(=O)CCCBr |
Specifications
| 4897-84-1 | |
| C5H9BrO2 | |
| methyl 4-bromobutyrate, 4-bromobutyric acid methyl ester, methyl-4-bromobutyrate, butanoic acid, 4-bromo-, methyl ester, methyl4-bromobutyrate, butyric acid, 4-bromo-, methyl ester, methyl gamma-bromobutyrate, acmc-1amn9, 4-brombutyric acid methyl ester, 4-bromobutyric acid methyl-ester | |
| COC(=O)CCCBr | |
| 181.03 | |
| 181.03 | |
| Liquid |
| Yellow | |
| MFCD00041482 | |
| QAWFLJGZSZIZHO-UHFFFAOYSA-N | |
| methyl 4-bromobutanoate | |
| 107604 | |
| ≥98.0% (GC) | |
| Methyl 4-Bromobutyrate |
Safety and Handling
TSCA : No