missing translation for 'onlineSavingsMsg'
Learn More

Methyl 4-Bromobutyrate 98.0+%, TCI America™

Catalog No. B230825G
Change view
Click to view available options
Quantity:
25 g
250 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B230825G 25 g
B2308250G 250 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B230825G Supplier TCI America Supplier No. B230825G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 4897-84-1
Molecular Formula C5H9BrO2
Molecular Weight (g/mol) 181.03
MDL Number MFCD00041482
InChI Key QAWFLJGZSZIZHO-UHFFFAOYSA-N
PubChem CID 107604
IUPAC Name methyl 4-bromobutanoate
SMILES COC(=O)CCCBr

Specifications

Color Yellow
Quantity 25 g
Synonym methyl 4-bromobutyrate, 4-bromobutyric acid methyl ester, methyl-4-bromobutyrate, butanoic acid, 4-bromo-, methyl ester, methyl4-bromobutyrate, butyric acid, 4-bromo-, methyl ester, methyl gamma-bromobutyrate, acmc-1amn9, 4-brombutyric acid methyl ester, 4-bromobutyric acid methyl-ester
Formula Weight 181.03
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material Methyl 4-Bromobutyrate