missing translation for 'onlineSavingsMsg'
Learn More

Methyl 4-Bromobutyrate 98.0+%, TCI America™

Supplier:  TCI America B2308250G

Quantity 250 g

 View more versions of this product

Catalog No. B2308250G


Only null left

Description

Description

Specifications

Chemical Identifiers

4897-84-1
181.03
QAWFLJGZSZIZHO-UHFFFAOYSA-N
methyl 4-bromobutanoate
C5H9BrO2
MFCD00041482
107604
COC(=O)CCCBr

Specifications

Methyl 4-Bromobutyrate
Yellow
250 g
methyl 4-bromobutyrate, 4-bromobutyric acid methyl ester, methyl-4-bromobutyrate, butanoic acid, 4-bromo-, methyl ester, methyl4-bromobutyrate, butyric acid, 4-bromo-, methyl ester, methyl gamma-bromobutyrate, acmc-1amn9, 4-brombutyric acid methyl ester, 4-bromobutyric acid methyl-ester
COC(=O)CCCBr
181.03
181.03
Liquid
4897-84-1
C5H9BrO2
MFCD00041482
QAWFLJGZSZIZHO-UHFFFAOYSA-N
methyl 4-bromobutanoate
107604
≥98.0% (GC)
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

TSCA : No

SDS
Documents

Documents

Promotions

Promotions