accessibility menu, dialog, popup

UserName

H3COOBr

CAS RN 4897-84-1

IUPAC Name: methyl 4-bromobutanoate
Synonyms: methyl 4-bromobutanoate
Molecular Weight (g/mol): 181.03
Molecular Formula: C5H9BrO2
InChi Key: QAWFLJGZSZIZHO-UHFFFAOYSA-N
SMILES: COC(=O)CCCBr
Percent Purity 
  • (2)
  • (2)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
Boiling Point 
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (4)
  • (2)
  • (4)
  • (2)
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (2)
  • (2)