missing translation for 'onlineSavingsMsg'
Learn More

Methyl 3,4-Dichlorophenylacetate 98.0+%, TCI America™

Numéro de catalogue. M25575G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Numéro de catalogue. Quantity
M25575G 5 g
M255725G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Numéro de catalogue. M25575G Fournisseur TCI America Code fournisseur M25575G
Il en reste null

Identifiants chimiques

CAS 6725-44-6
Molecular Formula C9H8Cl2O2
Molecular Weight (g/mol) 219.061
MDL Number MFCD00800680
InChI Key FWVOAIHAIWHHDM-UHFFFAOYSA-N
Synonym methyl 2-3,4-dichlorophenyl acetate, methyl 3,4-dichlorophenylacetate, 3,4-dichlorophenylacetic acid methyl ester, 3,4-dichloro-phenyl-acetic acid methyl ester, benzeneacetic acid, 3,4-dichloro-, methyl ester, pubchem20251, acmc-1bhwn, methyl 3,4-dichloro-phenylacetate, methyl 2-3,4-dichlorophenyl ethanoate, 3,4 dichlorophenyl acetic acid methyl ester
PubChem CID 1268257
IUPAC Name methyl 2-(3,4-dichlorophenyl)acetate
SMILES COC(=O)CC1=CC(=C(C=C1)Cl)Cl

Spécifications

Color Yellow
Quantity 5 g
Formula Weight 219.06
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material Methyl 3,4-Dichlorophenylacetate