missing translation for 'onlineSavingsMsg'
Learn More

meso-1,2-Dibromo-1,2-diphenylethane, 97%

Catalog No. AAL0495614
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL0495614 25 g
AAL0495606 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL0495614 Supplier Thermo Scientific Chemicals Supplier No. L0495614
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 13440-24-9 | C14H12Br2 | 340.058 g/mol

meso-1,2-Dibromo-1,2-diphenylethane is used to study the reaction of ± and meso-SBr2 with 9-substituted fluorenide ions in dimethyl sulfoxide. It undergoes electrochemical dehalogenation in acetonitrile.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
meso-1,2-Dibromo-1,2-diphenylethane is used to study the reaction of ± and meso-SBr2 with 9-substituted fluorenide ions in dimethyl sulfoxide. It undergoes electrochemical dehalogenation in acetonitrile.

Solubility
Insoluble in water.

Notes
Incompatible materials are Strong oxidizing agents. Store in cool place. Keep container tightly closed in a dry and well-ventilated place.

Chemical Identifiers

CAS 13440-24-9
Molecular Formula C14H12Br2
Molecular Weight (g/mol) 340.058
MDL Number MFCD00000137
InChI Key GKESIQQTGWVOLH-LSLKUGRBSA-N
Synonym meso-dibromostilbene
PubChem CID 102063597
IUPAC Name [(1S)-1,2-dibromo-2-phenylethyl]benzene
SMILES C1=CC=C(C=C1)C(C(C2=CC=CC=C2)Br)Br

Specifications

Melting Point ∽241°C (decomposition)
Quantity 25 g
UN Number UN1759
Beilstein 2050820
Solubility Information Insoluble in water.
Formula Weight 340.07
Percent Purity 97%
Chemical Name or Material meso-1,2-Dibromo-1,2-diphenylethane
Hazard Category H314-H335
Hazard Statement GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
Precautionary Statement P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c
DOTInformation Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: CORROSIVE SOLIDS, N.O.S.
EINECSNumber 227-324-7
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only