missing translation for 'onlineSavingsMsg'
Learn More

Mercury Standard For ICP, 10,000μg/mL in 10% HNO3, SPEX CertiPrep™

ISO/IEC Guide 34 Certified, NIST-Traceable Certified Reference Material

Supplier:  SPEX CertiPrep Group LLC PLHG43Y

 View more versions of this product

Catalog No. PLHG4-3Y


Only null left
Explore available promotions

Includes: NIST Traceable Certificate of Analysis accredited to ISO/IEC 17025 and ISO/IEC Guide 34, Safety Data Sheet

Description

Description

Inorganic Standard manufactured for AA, ICP, ICP/MS, IC, XRF, and other analytical instrumentation

AnalyteConcentration
Hg10,000μg/mL

Certifications

Accredited by A2LA for ISO/IEC 17025 and ISO/IEC Guide 34 and certified by UL-DQS to ISO 9001

Compliance

GHS/OSHA Complaint

Specifications

Specifications

Mercury Standard
Assurance Certificate of Analysis reports up to 30 elements at ppb levels.
Up to 68 elements are scanned with found values for ICP and ICP-MS standards. The found value of the trace metallic impurities is reported on the Certificate of Analysis.
MFCD00011035
AA, ICP, ICP/MS, IC, XRF, and Other Analytical Instrumentation
QSHDDOUJBYECFT-UHFFFAOYSA-N
10% HNO3
200.59
23931
Assurance
Triple-leached LDPE bottle
7439-97-6 , 7697-37-2 , 7732-18-5
Hg
AA, ICP
Certified Reference Material
[Hg]
mercury
125 mL
CHEBI:16170

Chemical Identifiers

7439-97-6
200.59
QSHDDOUJBYECFT-UHFFFAOYSA-N
CHEBI:16170
[Hg]
Hg
MFCD00011035
23931
mercury
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

Warning: Causes skin irritation, serious eye irritation Wear protective gloves/eye/face protection. If in eyes, rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing. If skin/eye irritation occurs, get medical advice/attention.

DOTInformation : Hazardous

Recommended Storage : Ambient

SDS Documents

Documents

Promotions

Promotions

Product intended for laboratory use only