missing translation for 'onlineSavingsMsg'
Learn More

Mercury Standard For ICP, 10,000μg/mL in 10% HNO3, SPEX CertiPrep™

ISO/IEC Guide 34 Certified, NIST-Traceable Certified Reference Material

$443.50 - $443.50

Chemical Identifiers

CAS 7439-97-6
Molecular Formula Hg
Molecular Weight (g/mol) 200.59
MDL Number MFCD00011035
InChI Key QSHDDOUJBYECFT-UHFFFAOYSA-N
PubChem CID 23931
ChEBI CHEBI:16170
IUPAC Name mercury
SMILES [Hg]
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
PLHG4-3Y
SDS View Documents
SPEX CertiPrep Group LLC
PLHG43Y
125 mL Triple-leached LDPE bottle
Each for $443.50
Only null left
 
Description

Description

Inorganic Standard manufactured for AA, ICP, ICP/MS, IC, XRF, and other analytical instrumentation

AnalyteConcentration
Hg10,000μg/mL

Certifications

Accredited by A2LA for ISO/IEC 17025 and ISO/IEC Guide 34 and certified by UL-DQS to ISO 9001

Compliance

GHS/OSHA Complaint

Specifications

Chemical Identifiers

7439-97-6
200.59
QSHDDOUJBYECFT-UHFFFAOYSA-N
CHEBI:16170
[Hg]
Hg
MFCD00011035
23931
mercury

Specifications

Triple-leached LDPE bottle
7439-97-6 , 7697-37-2 , 7732-18-5
Hg
AA, ICP
Certified Reference Material
[Hg]
mercury
125 mL
CHEBI:16170
Mercury Standard
Assurance Certificate of Analysis reports up to 30 elements at ppb levels.
Up to 68 elements are scanned with found values for ICP and ICP-MS standards. The found value of the trace metallic impurities is reported on the Certificate of Analysis.
MFCD00011035
AA, ICP, ICP/MS, IC, XRF, and Other Analytical Instrumentation
QSHDDOUJBYECFT-UHFFFAOYSA-N
10% HNO3
200.59
23931
Assurance
Videos
Safety and Handling

Safety and Handling

Warning: Causes skin irritation, serious eye irritation Wear protective gloves/eye/face protection. If in eyes, rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing. If skin/eye irritation occurs, get medical advice/attention.

DOTInformation : Hazardous

Recommended Storage : Ambient

SDS
Documents

Documents

Product intended for laboratory use only