missing translation for 'onlineSavingsMsg'
Learn More

Bulwark™ Men's Excel FR™ ComforTouch™ Grey Premium Flame Resistant Coverall

Catalog No. 17204450
Change view
Click to view available options
Size:
2X-Large (Long)
Large (Long)
X-Large (Long)
Small
Medium
Large
X-Large
2X-Large
3X-Large
9 product options available for selection
Product selection table with 9 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
17204450 2X-Large (Long)
17204051 Small
17204358 Medium
17204153 Large
17204284 X-Large
17203946 2X-Large
17204369 3X-Large
17204231 Large (Long)
17204033 X-Large (Long)
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 9 options available.
9 options
Catalog No. 17204450 Supplier Bulwark™ Supplier No. CLC8GY LN XXL

missing translation for 'validMainframeMsg'

Men's Excel FR™ ComforTouch™ Grey Premium Flame Resistant Coverall provides CAT 2 FR/AR protection and extra warmth for cold weather environments, without added bulk.

  • This premium coverall provides CAT 3 FR/AR protection and extra warmth for cold weather environments, without added bulk.
  • It also accommodates snap on FR hoods for additional insulation from the elements.
  • Certifications Level 1: ASTM F1506, NFPA 2112, NFPA 70E
  • Certifications Level 2: Category 3 Protection
  • Hazard: Flash Fire, Arc Flash
  • Primary Closure: Concealed Zipper
  • Wash Care: Industrial Laundry - Heavy Soil, Industrial Laundry - Light Soil, Home Wash
  • Fabric: Shell: Flame resistant, 7 oz. (235 g/m&superscript_2;) Twill, Water-repellent, Insulation Package: 14 oz. (474 g/m&superscript_2;)
  • Blend: Shell: 88% Cotton/ 12% Nylon, Lining: 100% Cotton, Insulation Package: 100% Modacrylic
  • Closure: Two-way, extra heavy-duty Nomex™ taped brass zipper front with concealed snap closures
  • Collar: Two-piece, insulated, stand-up collar with concealed snap closure
  • Pocket: Seven large patch pockets, two with hook-and-loop closure
  • Features: Full Nomex™ taped ankle-to-hip brass zippers, concealed snap closure
  • Product Collection: Excel FR Comfortouch
  • Protection: ATPV Arc Rating: 36
  • Country of Origin: Imported, Made in USA Fabric
  • Fit Details: Designed to be worn over other clothing. Fit over work boots.
  • Gender: Male
  • Materials: Cotton, Nylon
  • Brand: Bulwark™

Specifications

Product Type Flame-resistant Coveralls
Size 2X-Large (Long)
Gender Men's
Material Cotton, Nylon
Color Gray
Disposable Reusable
Waist Style Elastic (Stitched-in)
Ankle Style Open
Cuff Style Open
Neck Style Collared
Hood Type Unhooded
Closure Type Snap
Seam Type Stitched
Sterility Non-sterile
Protection Class CAT 3
Resistance Flame
Collar Type Two-Piece, Insulated, Stand-Up Collar
Arc Flash Rating 36
Certifications/Compliance ASTM F1506, NFPA 2112, NFPA 70E
No. of Pockets 7
Show More Show Less