Learn More
National Safety Apparel™ MD Energy Arc Goggle
Catalog No. 17001702
Encompass
Change view
Click to view available options
Black and blue
Catalog No.
17001702 Black and blue
1 options
Catalog No. 17001702 Supplier National Safety Apparel™ Supplier No. H08GGL
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Designed specifically to address the requirements of arc flash protection
  • The optically-correct hard-coated outer lens has an anti-fog inner lens baffled top and bottom vents
  • Special adjusters allow the elastic strap tension to be adjusted while wearing gloves
  • Goggle has an arc rating of 27 cal/cm²
  • Use in combination with the Carbon/Kevlar Balaclava Liner (#27181) for a lightweight cost-effective alternative to faceshields
Specifications
Size Medium
Material Plastic, Rubber
Color Blue
Seam Type Stitched
Arc Flash Rating 26 cal/cm²
Certifications/Compliance CAT 3, NFPA 70 E, ASTM F2178, and Z87+