missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Matrix™ SepraSeal and DuraSeal Capping Systems
GREENER_CHOICE

Accommodate a range of application and storage requirements. These flexible capping options allow automated piercing, visual identification and sterility preference.

Supplier:  Thermo Scientific™ 4465YEL

 Afficher plus de versions de ce produit

Numéro de catalogue. 04100549


Il en reste null

Description

Description

Accommodate a range of application and storage requirements. Thermo Scientific™ Matrix™ SepraSeal and DuraSeal tube sealing solutions complement the innovative line of Thermo Scientific Matrix 2D and non-2D coded storage tubes. Available in a range of formats to allow automated piercing, visual identification and sterility preference to meet your individual storage needs.

Sample Access Options

  • Designed for storage down to -20°C, Matrix SepraSeal mats are available in sterile, non-sterile, solid, or pre-split, and a range of colors for quick visual sample identification
  • Solid seals can be removed for sample access with the Thermo Scientific Matrix SepraSeal cap removal tool, or can be pierced once
  • For applications involving repetitive sample access, a pre-split Matrix SepraSeal can be pierced by automated liquid handling equipment, requiring very little pressure, while maintaining seal integrity with self-sealing closure
  • For applications requiring storage down to -80°C, permanently installed DuraSeals are an ultra-robust SepraSeal option—can also be used for repetitive piercing procedures
  • The Matrix DuraSeals Pre-Split V-Shaped design reliably and accurately guides automated liquid handling needles into the same location over multiple piercings—self-sealing design maintains sample integrity and prevents loss during storage

Application Flexibility

  • Elastomer ensures sealing performance will not degrade, even after multiple cap piercings
  • The Matrix SepraSeal tube sealing solution offers the flexibility of individual tube capping while also providing the ability to apply 96 caps at once
  • Supplied in mats of 96 caps each, sealing is secure and consistent every time when applied with the Thermo Scientific SuperSealer
  • After the SuperSealer applies the mat, simply pull up the backing sheet to reveal 96 individually sealed tubes—for applications where multiple piercings are required
  • The Matrix DuraSeal capping system is available in bulk or pre-installed in a wide array of our storage tubes
Spécifications

Identifiants chimiques

Croygenic Tube Closure

Spécifications

Matrix™ SepraSeal
Mats, Pre-Split
Pre-Split
Croygenic Tube Closure
Yellow
10 Mats of 96 Caps/Cs.
Non-sterile
missing translation for 'recommendations'

missing translation for 'recommendations'

Videos
FDS
Documents

Documents

Promotions

Promotions