missing translation for 'onlineSavingsMsg'
Learn More

Matrix Modifier, 1.0% Pd (10,000 ppm Pd), Ricca Chemical

Supplier:  Ricca Chemical Company MMPD10KN100

 View more versions of this product

Catalog No. MMPD10KN-100


Only null left

Description

Description

Specifications

Specifications

Matrix Modifier
88.0,9.23,0.88,0.16
91.0,9.61,0.91,0.16
Liquid
100 mL
Natural LDPE Bottle
1.0% Pd,10,000ppm Pd
7697-37-2 , 7440-05-3 , 7647-01-0
Brown
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions