missing translation for 'onlineSavingsMsg'
Learn More

Gibco™ Mammary Epithelial Growth Supplement (MEGS)

Catalog No. S0155
Encompass
Change view
Click to view available options
Quantity:
5 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
S0155 5 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S0155 Supplier Gibco™ Supplier No. S0155

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Mammary Epithelial Growth Supplement (MEGS)

A sterile, concentrated (100X) solution intended for use as one component in a complete culture environment for the growth of human mammary epithelial cells. Each 5 ml bottle of MEGS contains all of the growth factors, hormones, and tissue extracts necessary for the culture of mammary epithelial cells and is the correct amount of supplement for a 500 ml bottle of Medium 171.

When a 500 ml bottle of medium is supplemented with MEGS, the final concentrations of the components in the supplemented medium are:

  • Bovine pituitary extract (BPE) (0.4% v/v)
  • Recombinant human insulin-like growth factor-I (1μg/ml)
  • Hydrocortisone (0.5μg/ml)
  • Human epidermal growth factor (3 ng/ml)
  • Specifications

    Content And Storage Store at -20°C in a non-frost-free freezer.
    Cell Type Epithelial Cells (Mammary)
    Concentration 100 X
    Product Type Growth Supplement
    Sterility Sterile
    Product Line Cascade Biologics
    Quantity 5 mL
    Shipping Condition Dry Ice
    Species Human
    Is Mammary Epithelial Growth Supplement (MEGS) suitable for MCF10A culture?

    Yes, Mammary Epithelial Growth Supplement (MEGS) is suitable for MCF10A culture. You can also include 5% horse serum if desired.

    For Research Use Only. Not for use in diagnostic procedures.