missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Magnesium chloride hexahydrate, 99%, for biochemistry

Numéro de catalogue. p-4813527
Change view
Click to view available options
Quantity:
500 g
2.5 kg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Numéro de catalogue. Quantity
AC447155000 500 g
AC447150025 2.5 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Numéro de catalogue. AC447155000 Fournisseur Thermo Scientific Chemicals Code fournisseur 447155000
Il en reste null

CAS: 7791-18-6 | Cl2H12MgO6 | 203.30 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Identifiants chimiques

CAS 7791-18-6
Molecular Formula Cl2H12MgO6
Molecular Weight (g/mol) 203.30
MDL Number MFCD00149781
InChI Key DHRRIBDTHFBPNG-UHFFFAOYSA-L
IUPAC Name magnesium(2+) hexahydrate dichloride
SMILES O.O.O.O.O.O.[Mg++].[Cl-].[Cl-]

Specifications

Melting Point 117.0°C
Quantity 500 g
Assay Percent Range 98.5% min. (Complexometry)
Linear Formula MgCl2·6H2O
Merck Index 15,5726
Solubility Information Solubility in water: 2350g/L (20°C). Other solubilities: soluble in alcohol
Formula Weight 203.31
Percent Purity 99%
Grade Biochemical
Packaging Plastic bottle
Chemical Name or Material Magnesium chloride hexahydrate
Show More Show Less

RUO – Research Use Only