missing translation for 'onlineSavingsMsg'
Learn More

Bel Art™ Magic Touch 2™ Ice Pans

Superior insulating properties combined with high chemical resistance

Supplier:  Bel Art™ M168079101

 Afficher plus de versions de ce produit

Numéro de catalogue. 11999066


Get More Bel-Art Lab Products with Purchase
Afficher toutes les promotions
Il en reste null

Description

Description

Superior insulating properties combined with high chemical resistance

  • Compatible not only with wet ice, but also with ultra-low temperature materials including dry ice, salt slurries, dry ice solvent slurries, or liquid nitrogen
  • Able to withstand a wide temperature range of -196°C (-320°F) to 100°C (212°F)
  • High-quality, expanded urethane laboratory ice pans are lightweight, durable, non-sweating and impervious to moisture and odors
  • Lightly textured, non-slip finish and raised edge provides secure transport and a stylish look
  • Feature a convenient pour spout, drip-proof rims, a fill line indicator, and a snug-fitting, keyed lid and base for easy stacking
  • 1L pan does not include pour spout or fill line indicator
Spécifications

Identifiants chimiques

Lightly textured, Non-slip
Laboratory Ice Pan
Not Autoclavable
-196°C to +100°C
Urethane
24 Microtubes
No

Spécifications

20 cm
42 cm
30.3 cm
2.04 gal.
Lightly textured, Non-slip
Laboratory Ice Pan
9 L
Not Autoclavable
-196°C to +100°C
8 in.
16.5 in.
12 in.
Pour spout, drip-proof rims, fill line indicator, snug-fitting keyed Lid and base
Urethane
True Blue
24 Microtubes
No
missing translation for 'recommendations'

missing translation for 'recommendations'

Videos
FDS
Documents

Documents

Promotions

Promotions