Learn More
Bel Art™ Magic Touch 2™ Ice Pans
Superior insulating properties combined with high chemical resistance
$319.93 - $706.25
Specifications
| Design | Lightly textured, Non-slip |
|---|---|
| Material | Urethane |
| Type | Laboratory Ice Pan |
| Holds | 24 Microtubes |
| Autoclavable | Not Autoclavable |
| Catalog Number | Mfr. No. | Capacity (Metric) | Color | Includes | Length (Metric) | Price | Quantity & Availability | ||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Capacity (Metric) | Color | Includes | Length (Metric) | Price | Quantity & Availability | ||
|
11999050
|
Bel Art™
M168071101 |
1 L | True Blue | Pour spout, drip-proof rims, fill line indicator, snug-fitting keyed Lid and base | 18.3 cm |
Each for $319.93
|
|
||
|
11999068
|
Bel Art™
M168079103 |
9 L | Cherry Red | Pour spout, drip-proof rims, fill line indicator, snug-fitting keyed Lid and base | 42 cm |
Each for $672.18
|
|
||
|
11999066
|
Bel Art™
M168079101 |
9 L | True Blue | Pour spout, drip-proof rims, fill line indicator, snug-fitting keyed Lid and base | 42 cm |
Each for $706.18
|
|
||
|
11999067
|
Bel Art™
M168079102 |
9 L | Black Ebony | Pour spout, drip-proof rims, fill line indicator, snug-fitting keyed Lid and base | 42 cm |
Each for $703.50
|
|
||
|
11999065
|
Bel Art™
M168074104 |
4 L | Emerald Green | Pour spout, drip-proof rims, fill line indicator, snug-fitting keyed Lid and base | 30.4 cm |
Each for $511.04
|
|
||
|
11999051
|
Bel Art™
M168071102 |
1 L | Black Ebony | Pour spout, drip-proof rims, fill line indicator, snug-fitting keyed Lid and base | 18.3 cm |
Each for $320.07
|
|
||
|
11999069
|
Bel Art™
M168079104 |
9 L | Emerald Green | Pour spout, drip-proof rims, fill line indicator, snug-fitting keyed Lid and base | 42 cm |
Each for $706.25
|
|
||
|
11999053
|
Bel Art™
M168071104 |
1 L | Emerald Green | Pour spout, drip-proof rims, fill line indicator, snug-fitting keyed Lid and base | 18.3 cm |
Each for $320.21
|
|
||
|
11999062
|
Bel Art™
M168074101 |
4 L | True Blue | Pour spout, drip-proof rims, fill line indicator, snug-fitting keyed Lid and base | 30.4 cm |
Each for $512.75
|
|
||
|
11999063
|
Bel Art™
M168074102 |
4 L | Black Ebony | Pour spout, drip-proof rims, fill line indicator, snug-fitting keyed Lid and base | 30.4 cm |
Each for $474.07
|
|
||
|
11999064
|
Bel Art™
M168074103 |
4 L | Cherry Red | Pour spout, drip-proof rims, fill line indicator, snug-fitting keyed Lid and base | 30.4 cm |
Each for $512.75
|
|
||
|
11999052
|
Bel Art™
M168071103 |
1 L | Cherry Red | Pour spout, drip-proof rims, fill line indicator, snug-fitting keyed Lid and base | 18.3 cm |
Each for $320.21
|
|
||
Description
Superior insulating properties combined with high chemical resistance
- Compatible not only with wet ice, but also with ultra-low temperature materials including dry ice, salt slurries, dry ice solvent slurries, or liquid nitrogen
- Able to withstand a wide temperature range of -196°C (-320°F) to 100°C (212°F)
- High-quality, expanded urethane laboratory ice pans are lightweight, durable, non-sweating and impervious to moisture and odors
- Lightly textured, non-slip finish and raised edge provides secure transport and a stylish look
- Feature a convenient pour spout, drip-proof rims, a fill line indicator, and a snug-fitting, keyed lid and base for easy stacking
- 1L pan does not include pour spout or fill line indicator
Specifications
| Lightly textured, Non-slip | |
| Laboratory Ice Pan | |
| Not Autoclavable | |
| -196°C to +100°C |
| Urethane | |
| 24 Microtubes | |
| No |