missing translation for 'onlineSavingsMsg'
Learn More

MAbPac™ Capillary Reversed Phase HPLC Column

Catalog No. 03254130
Change view
Click to view available options
Length (Metric):
150 mm
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Length (Metric)
03254130 150 mm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 03254130 Supplier Thermo Fisher Scientific Supplier No. 164947

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Use this capillary column for high sensitivity HPLC or LC/MS characterization of intact proteins in top-down proteomics where sample amount is critically limited.

Use the Thermo Scientific™ MAbPac™ Capillary Reversed Phase HPLC Column high sensitivity HPLC or LC/MS characterization of intact proteins in top-down proteomics applications where sample amount is critically limited. This versatile column can be used for peptide-mapping characterization where the capillary format balances sensitivity with throughput. Additional applications are monoclonal antibodies (mAbs), fragments, variants, antibody drug conjugates (ADCs), PEGylated proteins, and bispecific proteins. These columns have excellent resolution, long column lifetime, and low carry-over.

Specifications

Column Format Capillary HPLC Column
Diameter (Metric) 0.15 mm
For Use With High sensitivity HPLC or LC-MS of intact proteins in top-down proteomics with limited sample
Length (Metric) 150 mm
Max. Pressure 8700 psi (600 bar)
Particle Shape N/A
Particle Size 4 μm
pH 0 to 9
Pore Size 1500 Å
Surface Area N/A
Temperature 110°C
USP Type N/A
Stationary Phase PSDVB (Polystyrene Divinylbenzene)
Product Line MAbPac
Packing Material PSDVB (Polystyrene Divinylbenzene)
Column Type Analytical HPLC Column
Endcapped N/A
Phase Reversed Phase
Show More Show Less