missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Lonza™ FlashGel™ System RNA Cassettes
Description
Contain pre-stained agarose gels and buffer, minimizing user exposure to potentially mutagenic stains and hazardous reagents
- Casettes fully enclose the gel, stain and running buffer
- Protect samples from contaminating RNases
- Designed for performance and purity, and are guaranteed RNase-free
- For research or manufacturing use only
- Not for use in in vitro diagnostic procedures, drug use, or for administration to humans or animals
Spécifications
Spécifications
| Concentration | 1.2% |
| Separation Range | 0.5 to 9 kb |
| Wells | 16 + 1-well |
| Formulation | 1.2% Agarose |
| For Use With (Application) | Analysis of total RNA, quick checks of native RNA and checking RNA degradation and mRNA purity |
| Kit Contents | Agarose Gel, Stain and Buffer |
| Material | Agarose |
| Quantity | 9 Pack |
| Length (English) Cassette | 4.5 in. |
| Length (English) Gel | 2.7 in. |
| Afficher plus de résultats |