missing translation for 'onlineSavingsMsg'
Learn More

Lithium standard solution, for AAS, 1 mg/ml Li in 2% HCl

CAS: 7439-93-2 | Li | 6.94 g/mol

$91.34 - $91.34

Specifications

Chemical Name or Material Lithium standard solution
Name Note 1 mg/ml Li in 2% HCl
Packaging Plastic bottle
CAS 7439-93-2 , 7647-01-0 , 7732-18-5
Density 0.53
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AC196081000
View Documents
Thermo Scientific Chemicals
196081000
100 mL
Each for $91.34
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

For atomic adsorption spectroscopy
Specifications

Specifications

Lithium standard solution
Plastic bottle
0.53
Li
MFCD00134051
[Li]
6.94
CHEBI:30145
AAS
1 mg/ml Li in 2% HCl
7439-93-2 , 7647-01-0 , 7732-18-5
Solution
995 to 1005 μg/mL of Li (1+) in 2% HCl (20°C)
WHXSMMKQMYFTQS-UHFFFAOYSA-N
lithium
3028194
6.94
Videos
SDS
Documents

Documents

RUO – Research Use Only