CAS RN 7439-93-2
Lithium foil, 1.5mm (0.06in) thick x 100mm (3.9in) wide, 99.9% (metals basis)
CAS: 7439-93-2 Molecular Formula: Li Molecular Weight (g/mol): 6.94 MDL Number: MFCD00134051 InChI Key: WHXSMMKQMYFTQS-UHFFFAOYSA-N Synonym: litio,litium,lithium, metallic,lithium, elemental,monohydride,unii-9fn79x2m3f,hsdb 647,hydride lih,hydrure de french,hsdb 549 PubChem CID: 3028194 ChEBI: CHEBI:30145 IUPAC Name: lithium SMILES: [Li]
Lithium wire, 3.2mm (0.125in) dia, 99.8%, Na 0.03% max
CAS: 7439-93-2 Molecular Formula: Li Molecular Weight (g/mol): 6.94 MDL Number: MFCD00134051 InChI Key: WHXSMMKQMYFTQS-UHFFFAOYSA-N Synonym: litio,litium,lithium, metallic,lithium, elemental,monohydride,unii-9fn79x2m3f,hsdb 647,hydride lih,hydrure de french,hsdb 549 PubChem CID: 3028194 ChEBI: CHEBI:30145 IUPAC Name: lithium SMILES: [Li]
Lithium granules, 1-6mm (0.04-0.2in), 99%
CAS: 7439-93-2 Molecular Formula: Li Molecular Weight (g/mol): 6.94 MDL Number: MFCD00134051 InChI Key: WHXSMMKQMYFTQS-UHFFFAOYSA-N Synonym: litio,litium,lithium, metallic,lithium, elemental,monohydride,unii-9fn79x2m3f,hsdb 647,hydride lih,hydrure de french,hsdb 549 PubChem CID: 3028194 ChEBI: CHEBI:30145 IUPAC Name: lithium SMILES: [Li]
Lithium, 99+%, granular, dry
CAS: 7439-93-2 Molecular Formula: Li Molecular Weight (g/mol): 6.94 MDL Number: MFCD00134051 InChI Key: WHXSMMKQMYFTQS-UHFFFAOYSA-N Synonym: litio,litium,lithium, metallic,lithium, elemental,monohydride,unii-9fn79x2m3f,hsdb 647,hydride lih,hydrure de french,hsdb 549 PubChem CID: 3028194 ChEBI: CHEBI:30145 IUPAC Name: lithium SMILES: [Li]
Lithium standard solution, for AAS, 1 mg/ml Li in 2% HCl
CAS: 7439-93-2 | Li | 6.94 g/mol
Lithium Standard For ICP, 1000μg/mL in 2% HNO3 Assurance Grade, SPEX CertiPrep™
CAS: 7439-93-2 Molecular Formula: Li Molecular Weight (g/mol): 6.94 MDL Number: MFCD00134051 InChI Key: WHXSMMKQMYFTQS-UHFFFAOYSA-N PubChem CID: 3028194 ChEBI: CHEBI:30145 IUPAC Name: lithium SMILES: [Li]
Lithium Standard For ICP, 10,000μg/mL in 5% HNO3, SPEX CertiPrep™
CAS: 7439-93-2 Molecular Formula: Li Molecular Weight (g/mol): 6.94 MDL Number: MFCD00134051 InChI Key: WHXSMMKQMYFTQS-UHFFFAOYSA-N PubChem CID: 3028194 ChEBI: CHEBI:30145 IUPAC Name: lithium SMILES: [Li]
Lithium Standard For ICP, 1000μg/mL in 2% HCl, SPEX CertiPrep™
CAS: 7439-93-2 Molecular Formula: Li Molecular Weight (g/mol): 6.94 MDL Number: MFCD00134051 InChI Key: WHXSMMKQMYFTQS-UHFFFAOYSA-N PubChem CID: 3028194 ChEBI: CHEBI:30145 IUPAC Name: lithium SMILES: [Li]
Lithium Single Element Ion Standard - Cation, SPEX CertiPrep™
CAS: 7439-93-2 Molecular Formula: Li Molecular Weight (g/mol): 6.94 MDL Number: MFCD00134051 InChI Key: WHXSMMKQMYFTQS-UHFFFAOYSA-N PubChem CID: 3028194 ChEBI: CHEBI:30145 IUPAC Name: lithium SMILES: [Li]
Lithium Standard for ICP/MS, 2% HNO3, SPEX CertiPrep™
CAS: 7439-93-2 Molecular Formula: Li Molecular Weight (g/mol): 6.94 MDL Number: MFCD00134051 InChI Key: WHXSMMKQMYFTQS-UHFFFAOYSA-N PubChem CID: 3028194 ChEBI: CHEBI:30145 IUPAC Name: lithium SMILES: [Li]