missing translation for 'onlineSavingsMsg'
Learn More

Lithium Standard for ICP/MS, 2% HNO3, SPEX CertiPrep™

ISO/IEC Guide 34 Certified, NIST-Traceable Certified Reference Material

Supplier:  SPEX CertiPrep Group LLC CLLI21BY

Catalog No. CLLI21BY

Only null left

Includes: NIST Traceable Certificate of Analysis accredited to ISO/IEC 17025 and ISO/IEC Guide 34, Safety Data Sheet

Description

Description

Inorganic Standard manufactured for AA, ICP, ICP/MS, IC, XRF, and other analytical instrumentation

AnalyteConcentration
Li1μg/mL

Certifications

Accredited by A2LA for ISO/IEC 17025 and ISO/IEC Guide 34 and certified by UL-DQS to ISO 9001

Compliance

GHS/OSHA Complaint

Specifications

Specifications

Lithium Standard
Triple-leached LDPE bottle
Up to 68 elements are scanned with found values for ICP and ICP-MS standards. The found value of the trace metallic impurities is reported on the Certificate of Analysis.
1.01006 g/cm3 at 20°C
Li
ICP-MS
Standard Solution
WHXSMMKQMYFTQS-UHFFFAOYSA-N
2% HNO3
6.94
3028194
Claritas PPT
ICP/MS
7439-93-2 , 7697-37-2 , 7732-18-5
2.0,0.0,98.0
Liquid
MFCD00134051
AA, ICP, ICP/MS, IC, XRF, and Other Analytical Instrumentation
ICP/ICP-MS Standard
[Li]
lithium
125 mL
CHEBI:30145
1mg/L

Chemical Identifiers

7439-93-2
6.94
WHXSMMKQMYFTQS-UHFFFAOYSA-N
CHEBI:30145
[Li]
Li
MFCD00134051
3028194
lithium
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

Causes skin irritation, serious eye irritation

DOTInformation : Hazardous

Recommended Storage : Ambient

SDS
Documents

Documents

Promotions

Promotions

Product intended for laboratory use only