missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Lithium AA Standard, 1000 ppm Li in 3% HNO3, Ricca Chemical
$112.43 - $112.43
Chemical Identifiers
| CAS | 554-13-2 |
|---|---|
| Molecular Formula | CLi2O3 |
| Molecular Weight (g/mol) | 73.89 |
| MDL Number | MFCD00011084 |
| InChI Key | XGZVUEUWXADBQD-UHFFFAOYSA-L |
| PubChem CID | 11125 |
| ChEBI | CHEBI:6504 |
| IUPAC Name | dilithium(1+) carbonate |
| SMILES | [Li+].[Li+].[O-]C([O-])=O |
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|
ALI1KN-100
|
Ricca Chemical Company
ALI1KN100 |
100 mL | Natural LDPE Bottle |
Each for $112.43
|
|
||||
Chemical Identifiers
| 554-13-2 | |
| 73.89 | |
| XGZVUEUWXADBQD-UHFFFAOYSA-L | |
| CHEBI:6504 | |
| [Li+].[Li+].[O-]C([O-])=O |
| CLi2O3 | |
| MFCD00011084 | |
| 11125 | |
| dilithium(1+) carbonate |
Specifications
| Natural LDPE Bottle | |
| 1,000ppm Li | |
| 96.0,5.62,0.51 | |
| Liquid | |
| MFCD00011084 | |
| < 2 | |
| XGZVUEUWXADBQD-UHFFFAOYSA-L | |
| dilithium(1+) carbonate | |
| 100 mL | |
| CHEBI:6504 |