missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Lithium AA Standard, 1000 ppm Li in 3% HNO3, Ricca Chemical
Supplier: Ricca Chemical Company ALI1KN100
Specifications
| Lithium AA Standard | |
| 554-13-2 , 7697-37-2 , 7732-18-5 | |
| 92.0,5.4,0.49 | |
| 1 mL = 1 mg Li | |
| CLi2O3 | |
| Colorless | |
| Approximately 0°C | |
| [Li+].[Li+].[O-]C([O-])=O | |
| 73.89 | |
| 11125 |
| Natural LDPE Bottle | |
| 1,000ppm Li | |
| 96.0,5.62,0.51 | |
| Liquid | |
| MFCD00011084 | |
| < 2 | |
| XGZVUEUWXADBQD-UHFFFAOYSA-L | |
| dilithium(1+) carbonate | |
| 100 mL | |
| CHEBI:6504 |
Chemical Identifiers
| 554-13-2 | |
| 73.89 | |
| XGZVUEUWXADBQD-UHFFFAOYSA-L | |
| CHEBI:6504 | |
| [Li+].[Li+].[O-]C([O-])=O |
| CLi2O3 | |
| MFCD00011084 | |
| 11125 | |
| dilithium(1+) carbonate |