missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Leupeptin Hemisulfate

Numéro de catalogue. p-7042046
Change view
Click to view available options
Quantity:
25 mg
100 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Numéro de catalogue. Quantity
AAJ61188MB 25 mg
AAJ61188MC 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Numéro de catalogue. AAJ61188MB Fournisseur Thermo Scientific Chemicals Code fournisseur J61188MB
Il en reste null

Reversible protease inhibitor which inhibits cathepsin B, calpain and trypsin | CAS: 103476-89-7 | C20H39N6O4+

Reversible protease inhibitor, which inhibits cathepsin B, calpain and trypsinPharmaceutical composition containing leupeptin hemisulfate is used as an anti-malarial agent. It is employed in the treatment of noise-induced hearing loss. It also protects the heart from myocardial stunning. Further, it is used to inhibit serine, cysteine proteases, plasmin, trypsin, papain, kallikrein and cathepsin B.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Reversible protease inhibitor, which inhibits cathepsin B, calpain and trypsinPharmaceutical composition containing leupeptin hemisulfate is used as an anti-malarial agent. It is employed in the treatment of noise-induced hearing loss. It also protects the heart from myocardial stunning. Further, it is used to inhibit serine, cysteine proteases, plasmin, trypsin, papain, kallikrein and cathepsin B.

Solubility
Soluble in water, ethanol, dimethyl formamide and acetic acid.

Notes
Store in a cool place. Incompatible with strong acids and strong bases.

Identifiants chimiques

CAS 103476-89-7
Molecular Formula C20H39N6O4+

Spécifications

Color White
Product Type Leupeptin
Formula Weight 475.59
Solubility Soluble in water at 50mg/ml
Content And Storage -30°C to -10°C
Form Powder
Quantity 25 mg
Recommended Storage Store at -20°C

RUO – Research Use Only