missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Leupeptin Hemisulfate

Reversible protease inhibitor which inhibits cathepsin B, calpain and trypsin | CAS: 103476-89-7 | C20H39N6O4+

Supplier:  Thermo Scientific Chemicals J61188MC

 View more versions of this product

Catalog No. AAJ61188MC


Only null left
Explore available promotions

Description

Description

Reversible protease inhibitor, which inhibits cathepsin B, calpain and trypsinPharmaceutical composition containing leupeptin hemisulfate is used as an anti-malarial agent. It is employed in the treatment of noise-induced hearing loss. It also protects the heart from myocardial stunning. Further, it is used to inhibit serine, cysteine proteases, plasmin, trypsin, papain, kallikrein and cathepsin B.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Reversible protease inhibitor, which inhibits cathepsin B, calpain and trypsinPharmaceutical composition containing leupeptin hemisulfate is used as an anti-malarial agent. It is employed in the treatment of noise-induced hearing loss. It also protects the heart from myocardial stunning. Further, it is used to inhibit serine, cysteine proteases, plasmin, trypsin, papain, kallikrein and cathepsin B.

Solubility
Soluble in water, ethanol, dimethyl formamide and acetic acid.

Notes
Store in a cool place. Incompatible with strong acids and strong bases.
Specifications

Specifications

White
103476-89-7
475.59
-30°C to -10°C
100 mg
Leupeptin
C20H39N6O4+
Soluble in water at 50mg/ml
Powder

Chemical Identifiers

103476-89-7
C20H39N6O4+
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

Recommended Storage : Store at -20°C

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only