missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific Chemicals Leupeptin, 90%, synthetic
CAS: 103476-89-7 | 0·5 H2O4S
Supplier: Thermo Scientific Chemicals 328350250
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
| Leupeptin | |
| Leupeptin | |
| 0·5 H2O4S | |
| Solubility in water: soluble in water | |
| 25 mg |
| White | |
| 103476-89-7 | |
| 475.6 | |
| Powder | |
| 90% |
Chemical Identifiers
| 103476-89-7 | |
| 0·5 H2O4S |
RUO – Research Use Only