missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Leupeptin, 90%, synthetic

Catalog No. p-4391382
Change view
Click to view available options
Quantity:
5 mg
25 mg
100 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC328350050 5 mg
AC328350250 25 mg
AC328351000 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AC328350050 Supplier Thermo Scientific Chemicals Supplier No. 328350050
Only null left

CAS: 103476-89-7 | 0·5 H2O4S

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 103476-89-7
Molecular Formula 0·5 H2O4S

Specifications

Color White
Product Type Leupeptin
Formula Weight 475.6
Solubility Solubility in water: soluble in water
Form Powder
Quantity 5 mg
Percent Purity 90%
Chemical Name or Material Leupeptin

RUO – Research Use Only