missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Lead Standard, 10 ppm Pb, Ricca Chemical
$402.83 - $402.83
Chemical Identifiers
| CAS | 7439-92-1 |
|---|---|
| Molecular Formula | Pb |
| Molecular Weight (g/mol) | 207.20 |
| MDL Number | MFCD00134050 |
| InChI Key | WABPQHHGFIMREM-UHFFFAOYSA-N |
| PubChem CID | 5352425 |
| ChEBI | CHEBI:27889 |
| IUPAC Name | lead |
| SMILES | [Pb] |
Chemical Identifiers
| 7439-92-1 | |
| 207.20 | |
| WABPQHHGFIMREM-UHFFFAOYSA-N | |
| CHEBI:27889 | |
| [Pb] |
| Pb | |
| MFCD00134050 | |
| 5352425 | |
| lead |
Specifications
| Natural Poly Bottle | |
| 10ppm Pb | |
| 100.0,1.35,0.05,0.0010 | |
| Pb | |
| Colorless | |
| WABPQHHGFIMREM-UHFFFAOYSA-N | |
| lead | |
| 500 mL | |
| CHEBI:27889 |