missing translation for 'onlineSavingsMsg'
Learn More

Lead Standard, 10 ppm Pb, Ricca Chemical

Catalog No. 429716
Change view
Click to view available options
Quantity:
500 mL
Packaging:
Natural Poly Bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
429716 500 mL Natural Poly Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 429716 Supplier Ricca Chemical Company Supplier No. 429716
Only null left

Chemical Identifiers

CAS 7439-92-1 , 7697-37-2 , 7697-37-2 , 7732-18-5
Molecular Formula Pb
Molecular Weight (g/mol) 207.20
MDL Number MFCD00134050
InChI Key WABPQHHGFIMREM-UHFFFAOYSA-N
PubChem CID 5352425
ChEBI CHEBI:27889
IUPAC Name lead
SMILES [Pb]

Specifications

Packaging Natural Poly Bottle
Concentration or Composition Notes 10ppm Pb
CAS Min % 97.0, 1.29, 0.05, 0.0010
CAS Max % 100.0, 1.35, 0.05, 0.0010
Physical Form Liquid
Color Colorless
Boiling Point 100°C
Quantity 500 mL
Chemical Name or Material Lead Standard